siRNA Details
| Gene Symbol: Fbxw11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010033.1 | - | 1 | 340 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB730861.1 | - | 1 | 101 | Chr. 11 - 32680371 - 32680831 [+] on Build GRCm38 | |

Fbxw11
Genomic Map