siRNA Details
| Gene Symbol: 2210420L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019073.1 | - | 1 | 225 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708875.1 | - | 1 | 225 | Chr. 10 - 87571441 - 87571768 [+] on Build GRCm38 | |

2210420L05Rik
Genomic Map