siRNA Details
| Gene Symbol: Gm13391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096394.1 | - | 272 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AW913196.1 | - | 2 | 263 | Chr. 2 - 6472272 - 6472903 [+] on Build GRCm38 | |

Gm13391
Genomic Map