siRNA Details
| Gene Symbol: Gm32455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133388.1 | - | 458 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB616963.1 | - | 4 | 458 | Chr. 14 - 84048054 - 84075290 [-] on Build GRCm38 | |

Gm32455
Genomic Map