siRNA Details
| Gene Symbol: Atxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA435241.1 | - | 1 | 244 | ||
| AI642908.1 | - | 1 | 249 | |||
| AK019314.1 | - | 1 | 407 | |||
| AK135987.1 | - | 1 | 1912 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atxn1| Gene Symbol: Atxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA435241.1 | - | 1 | 244 | ||
| AI642908.1 | - | 1 | 249 | |||
| AK019314.1 | - | 1 | 407 | |||
| AK135987.1 | - | 1 | 1912 | |||