siRNA Details
| Gene Symbol: Atxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019314.1 | - | 1 | 30 | ||
| AK135987.1 | - | 1 | 1535 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712500.1 | - | 1 | 30 | Chr. 13 - 45552673 - 45553124 [-] on Build GRCm38 | |

Atxn1
Genomic Map