siRNA Details
| Gene Symbol: Gm11998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041795.1 | - | 3 | 599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11998| Gene Symbol: Gm11998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041795.1 | - | 3 | 599 | ||