siRNA Details
| Gene Symbol: Gm29687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139823.1 | - | 1 | 48 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725917.1 | - | 1 | 48 | Chr. 12 - 26092182 - 26210505 [-] on Build GRCm38 | |

Gm29687
Genomic Map