siRNA Details
| Gene Symbol: Gm16793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040376.1 | 3 | 1465 | |||
| GenBank mRNA | AA250323.1 | - | 1 | 44 | ||
| AK040224.1 | - | 3 | 1418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16793| Gene Symbol: Gm16793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040376.1 | 3 | 1465 | |||
| GenBank mRNA | AA250323.1 | - | 1 | 44 | ||
| AK040224.1 | - | 3 | 1418 | |||