siRNA Details
| Gene Symbol: Gm29678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085057.1 | - | 3 | 601 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB485359.2 | - | 1 | 138 | Chr. 1 - 190416475 - 190417145 [+] on Build GRCm38 | |

Gm29678
Genomic Map