siRNA Details
| Gene Symbol: E330013P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK027925.1 | - | 1 | 454 | ||
| AK036852.1 | - | 1 | 566 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB142958.2 | - | 1 | 45 | Chr. 19 - 60145208 - 60145880 [+] on Build GRCm38 | |

E330013P04Rik
Genomic Map