siRNA Details
| Gene Symbol: Cacna1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142841.1 | - | 1 | 2724 | ||
| BC094579.1 | - | 2 | 2654 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna1c| Gene Symbol: Cacna1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142841.1 | - | 1 | 2724 | ||
| BC094579.1 | - | 2 | 2654 | |||