siRNA Details
| Gene Symbol: Gm35288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029654.1 | - | 422 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615156.1 | - | 2 | 419 | Chr. 9 - 62303661 - 62324820 [+] on Build GRCm38 | |

Gm35288
Genomic Map