siRNA Details
| Gene Symbol: A330033J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102303.1 | 3 | 1769 | |||
| GenBank mRNA | AK034282.1 | - | 3 | 1769 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330033J07Rik| Gene Symbol: A330033J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102303.1 | 3 | 1769 | |||
| GenBank mRNA | AK034282.1 | - | 3 | 1769 | ||