siRNA Details
| Gene Symbol: Gm13580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046065.1 | 1 | 12 | |||
| GenBank mRNA | BC051520.1 | - | 1 | 21 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13580| Gene Symbol: Gm13580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046065.1 | 1 | 12 | |||
| GenBank mRNA | BC051520.1 | - | 1 | 21 | ||