siRNA Details
Gene Symbol: Gm13580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046065.1 | 3 | 479 | |||
GenBank mRNA | BC051520.1 | - | 3 | 488 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI158402.1 | - | 3 | 434 | Chr. 2 - 60411569 - 60412519 [+] on Build GRCm38 |
