siRNA Details
| Gene Symbol: LOC105246071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134561.1 | - | 2055 | |||
| Gene Symbol: AA589531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA589531.1 | - | 1 | 34 | ||

LOC105246071
Genomic Map