siRNA Details
| Gene Symbol: 2900074G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013781.1 | - | 1 | 309 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV155169.1 | - | 1 | 43 | Chr. 10 - 108774001 - 108774280 [-] on Build GRCm38 | |

2900074G08Rik
Genomic Map