siRNA Details
| Gene Symbol: Gm9806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015803.1 | - | 1 | 699 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU937106.1 | - | 1 | 106 | Chr. 19 - 16781141 - 16781908 [+] on Build GRCm38 | |

Gm9806
Genomic Map