siRNA Details
| Gene Symbol: Cct4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011873.1 | - | 1279 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK335126.1 | - | 1 | 463 | Chr. 11 - 23005044 - 23005651 [+] on Build GRCm38 | |

Cct4
Genomic Map