siRNA Details
| Gene Symbol: Gm17102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK163099.1 | - | 2 | 845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17102| Gene Symbol: Gm17102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK163099.1 | - | 2 | 845 | ||