siRNA Details
| Gene Symbol: Gm17102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK163099.1 | - | 1 | 35 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724782.1 | - | 1 | 35 | Chr. 7 - 41393698 - 41397243 [+] on Build GRCm38 | |

Gm17102
Genomic Map