siRNA Details
| Gene Symbol: Mob1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042552.1 | - | 2138 | |||
| AK048590.1 | - | 448 | ||||
| AK051056.1 | - | 1111 | ||||
| AK163091.1 | - | 1335 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mob1b| Gene Symbol: Mob1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042552.1 | - | 2138 | |||
| AK048590.1 | - | 448 | ||||
| AK051056.1 | - | 1111 | ||||
| AK163091.1 | - | 1335 | ||||