siRNA Details
| Gene Symbol: Gm7715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131765.1 | - | 304 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707175.1 | - | 1 | 304 | Chr. 14 - 32513286 - 32513752 [-] on Build GRCm38 | |

Gm7715
Genomic Map