siRNA Details
| Gene Symbol: 4631402F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014506.1 | - | 1 | 391 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713919.1 | - | 1 | 391 | Chr. 17 - 65871467 - 65872418 [-] on Build GRCm38 | |

4631402F24Rik
Genomic Map