siRNA Details
| Gene Symbol: 4631402F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014506.1 | - | 1 | 956 | ||
| AV228829.1 | - | 1 | 224 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4631402F24Rik| Gene Symbol: 4631402F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014506.1 | - | 1 | 956 | ||
| AV228829.1 | - | 1 | 224 | |||