siRNA Details
| Gene Symbol: Acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077294.1 | - | 1 | 424 | ||
| AK080187.1 | - | 1 | 972 | |||
| AK137170.1 | - | 1 | 980 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acta2| Gene Symbol: Acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077294.1 | - | 1 | 424 | ||
| AK080187.1 | - | 1 | 972 | |||
| AK137170.1 | - | 1 | 980 | |||