siRNA Details
Gene Symbol: Acta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077294.1 | - | 1 | 206 | ||
AK080187.1 | - | 1 | 760 | |||
AK137170.1 | - | 1 | 767 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Acta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077294.1 | - | 1 | 206 | ||
AK080187.1 | - | 1 | 760 | |||
AK137170.1 | - | 1 | 767 |