siRNA Details
| Gene Symbol: 2210406O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144870.1 | - | 3 | 958 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB818630.1 | - | 2 | 236 | Chr. 5 - 36447611 - 36455997 [-] on Build GRCm38 | |

2210406O10Rik
Genomic Map