siRNA Details
| Gene Symbol: A930005K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080704.1 | - | 2 | 1276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930005K07Rik| Gene Symbol: A930005K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080704.1 | - | 2 | 1276 | ||