siRNA Details
| Gene Symbol: Gm20110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038019.1 | 3 | 1012 | |||
| GenBank mRNA | AK039362.1 | - | 3 | 1013 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20110| Gene Symbol: Gm20110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038019.1 | 3 | 1012 | |||
| GenBank mRNA | AK039362.1 | - | 3 | 1013 | ||