siRNA Details
Gene Symbol: AI314278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102276.1 | 2 | 308 | |||
GenBank mRNA | AK143758.1 | - | 2 | 306 | ||
AK165495.1 | - | 2 | 309 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI314278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102276.1 | 2 | 308 | |||
GenBank mRNA | AK143758.1 | - | 2 | 306 | ||
AK165495.1 | - | 2 | 309 |