siRNA Details
Gene Symbol: AI314278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102276.1 | 1 | 87 | |||
GenBank mRNA | AK143758.1 | - | 1 | 85 | ||
AK165495.1 | - | 1 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI314278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102276.1 | 1 | 87 | |||
GenBank mRNA | AK143758.1 | - | 1 | 85 | ||
AK165495.1 | - | 1 | 88 |