siRNA Details
| Gene Symbol: AI314278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102276.1 | 2 | 239 | |||
| GenBank mRNA | AK143758.1 | - | 2 | 237 | ||
| AK165495.1 | - | 2 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI314278| Gene Symbol: AI314278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102276.1 | 2 | 239 | |||
| GenBank mRNA | AK143758.1 | - | 2 | 237 | ||
| AK165495.1 | - | 2 | 240 | |||