siRNA Details
| Gene Symbol: A930027M08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020897.1 | - | 1 | 69 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742262.1 | - | 1 | 67 | Chr. 12 - 75820288 - 75820757 [+] on Build GRCm38 | |

A930027M08Rik
Genomic Map