siRNA Details
| Gene Symbol: Gm26544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033312.1 | - | 3 | 376 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB624477.1 | - | 3 | 373 | Chr. 3 - 116150481 - 116163329 [+] on Build GRCm38 | |

Gm26544
Genomic Map