siRNA Details
| Gene Symbol: 3830422I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014451.1 | - | 1 | 831 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3830422I06Rik| Gene Symbol: 3830422I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014451.1 | - | 1 | 831 | ||