siRNA Details
| Gene Symbol: AW822252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110489.1 | 6 | 841 | |||
| GenBank mRNA | AK082553.1 | - | 6 | 842 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB389450.2 | - | 4 | 377 | Chr. X - 53696922 - 53715404 [-] on Build GRCm38 | |

AW822252
Genomic Map