siRNA Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009395.1 | - | 1 | 977 | ||
| AK046354.1 | - | 1 | 1018 | |||
| BC023871.1 | - | 1 | 762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210018M11Rik| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009395.1 | - | 1 | 977 | ||
| AK046354.1 | - | 1 | 1018 | |||
| BC023871.1 | - | 1 | 762 | |||