siRNA Details
| Gene Symbol: Clgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015491.1 | - | 1 | 422 | ||
| AK034525.1 | - | 15 | 3323 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715039.1 | - | 1 | 420 | Chr. 8 - 83427552 - 83428343 [+] on Build GRCm38 | |

Clgn
Genomic Map