siRNA Details
| Gene Symbol: 2610034E01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011660.1 | - | 1 | 976 | ||
| AK142273.1 | - | 1 | 774 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610034E01Rik| Gene Symbol: 2610034E01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011660.1 | - | 1 | 976 | ||
| AK142273.1 | - | 1 | 774 | |||