siRNA Details
| Gene Symbol: D5Ertd615e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015324.1 | - | 1 | 35 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714879.1 | - | 1 | 35 | Chr. 5 - 45111667 - 45175693 [-] on Build GRCm38 | |

D5Ertd615e
Genomic Map