siRNA Details
| Gene Symbol: Gm28703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043331.1 | - | 2 | 213 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640798.1 | - | 2 | 213 | Chr. 9 - 90825784 - 90828860 [-] on Build GRCm38 | |

Gm28703
Genomic Map