siRNA Details
| Gene Symbol: 4930588G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016366.1 | - | 4 | 1205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930588G17Rik| Gene Symbol: 4930588G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016366.1 | - | 4 | 1205 | ||