siRNA Details
| Gene Symbol: 4930588G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016366.1 | - | 3 | 658 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716013.1 | - | 3 | 658 | Chr. 7 - 120670303 - 120675017 [+] on Build GRCm38 | |

4930588G17Rik
Genomic Map