siRNA Details
| Gene Symbol: Gm26562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132624.1 | - | 2 | 516 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714286.1 | - | 2 | 516 | Chr. 16 - 30202285 - 30205879 [-] on Build GRCm38 | |

Gm26562
Genomic Map