siRNA Details
| Gene Symbol: 4930587E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016361.1 | - | 5 | 675 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716007.1 | - | 5 | 676 | Chr. 6 - 102266121 - 102336677 [+] on Build GRCm38 | |

4930587E11Rik
Genomic Map