siRNA Details
| Gene Symbol: Teddm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017892.1 | - | 644 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717725.1 | - | 1 | 643 | Chr. 1 - 153899313 - 153900228 [-] on Build GRCm38 | |

Teddm2
Genomic Map