siRNA Details
| Gene Symbol: Teddm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017892.1 | - | 885 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV312559.1 | - | 1 | 168 | Chr. 1 - 153899313 - 153899512 [-] on Build GRCm38 | |

Teddm2
Genomic Map