siRNA Details
Gene Symbol: D830032E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102307.1 | 2 | 124 | |||
GenBank mRNA | AK081398.1 | - | 32 | |||
AK085943.1 | - | 2 | 99 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D830032E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102307.1 | 2 | 124 | |||
GenBank mRNA | AK081398.1 | - | 32 | |||
AK085943.1 | - | 2 | 99 |